Would you like to log in again?
Z-Leu-Val-Gly Diazomethyl Ketone
Supplier: Thermo Scientific
J64731-MB J64731-MB
AAJ64731-MBEA 461.59 USD InStock
AAJ64731-MB CAAAJ64731-MB
Z-Leu-Val-Gly Diazomethyl Ketone
Proteins and Peptides
Mimics part of the human cysteine proteinase-binding center; an irreversible inhibitor of human cysteine proteinase.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty