Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

8-Bromo-2'-deoxyadenosine 99%

Supplier: Thermo Scientific Chemicals
Warning

J65357-03 J65357-03
AAJ65357-03EA 163.31 USD InStock
AAJ65357-03 CAAAJ65357-03
8-Bromo-2'-deoxyadenosine 99%
8-Bromo-2'-deoxyadenosine
Powder


Formula: C₁₀H₁₂BrN₅O₃
MW: 330.142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01630970
CAS Number: 14985-44-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty