Would you like to log in again?
Casein Sodium Salt
Supplier: Thermo Scientific
J65590-A7 J65590-A7
AAJ65590-A7EA 238.36 USD InStock
AAJ65590-A7 CAAAJ65590-A7
Casein Sodium Salt
Proteins and Peptides
Casein is frequently used as a blocking agent in immunochemistry experiments. It is also used in the recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty