Would you like to log in again?
SP Bel-Art Tiered Test Tube Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
H18862-0000
71001-744CS 791.49 USD InStock
71001-744
SP Bel-Art Tiered Test Tube Rack, Bel-Art Products, a part of SP
Racks Tube Racks
Heavy-duty polypropylene rack is perfect for organization, storage, and transport of test tubes.
  • Heavy-duty design
  • Polypropylene rack construction
  • Chemically resistant
  • Autoclavable
The 16-place rack holds tubes 10–13 mm in diameter. Heat and chemical resistant, it is autoclavable at 121°C (250°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty