Would you like to log in again?
Still Head, Short Path, Ace Glass
Supplier: Ace Glass
9311-15
89074-342EA 409.91 USD InStock
89074-342
Still Head, Short Path, Ace Glass
Distillation Components Distillation Heads
This jacketed distillation head is used for distilling samples up to 3mL
The jacket allows positioning of a coolant, such as dry ice/acetone, at the site of the desired condensation, thus reducing loss of valuable sample. Distillate can be removed with a Pasteur pipet. The head has an outer [ST] 14/20 joint at top and inner [ST] 14/20 joint at bottom. Fabricated for borosilicate glass.
Ordering information: Related items (available separately) for a complete short path still include: 89074-322 jacketed Liebig condenser with [ST] 14/20 joints; 89051-102 thermometer adapter, #7 Ace-Thred — [ST] 14/20 joint; and 89074-912 pear-shaped 5mL flask with ground [ST] 14/20 outer joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty