Would you like to log in again?
Corning® CMRL 1066 without L-glutamine, Corning
Supplier: Corning
Corning®
15-110-CV 15-110-CV
45001-114PK 371.93 USD InStock
45001-114 CA45001-114
Corning® CMRL 1066 without L-glutamine, Corning
Cell Culture Media
A less complex and extensively modified version of Medium 199, CMRL-1066 was designed initially for use with L-strain cells in unsupplemented culture
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
It has been used to maintain monolayer growth of permanent cell lines for years without protein supplementation. Although developed for use in serum-free cell culture, CMRL-1066 can be supplemented with serum and used to support the growth of many types of cells.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty