Would you like to log in again?
Flange, Conical, Ace Glass
Supplier: Ace Glass
15305-20
89078-282EA 392.49 USD InStock
89078-282
Flange, Conical, Ace Glass
Fittings and Connectors
This conical-type flange used by glassblowers to fabricate reaction flasks.
Supplied on medium-wall tubing, it uses a 89055-014 clamp (not supplied). Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty