Would you like to log in again?
Separatory Funnel Type Flask, Ace Glass Incorporated
Supplier: Ace Glass
7011-254 7011-258 7011-259 7011-255 7011-257 7011-261 7011-251 7011-244 7011-253 7011-256 7011-260 7011-249 7011-248 7011-252 7011-245 7011-262 7011-247 7011-246
89057-450EA 763.8 USD InStock
89057-450 89057-460 89057-454 89057-432 89057-442 89057-452 89057-430 89057-440 89057-462 89057-458 89057-436 89057-446 89057-456 89057-434 89057-444 89057-428 89057-438 89057-448
Separatory Funnel Type Flask, Ace Glass Incorporated
Funnels Separating Funnels
This separatory funnel type flask has three reinforced necks and a PTFE plug (6mm bore on 5000 mL, 4mm bore on all other sizes) stopcock bottom outlet
Fabricated with heavy walls, approximately 30% heavier than standard wall flasks. Fabricated from borosilicate glass.
Ordering information: For replacement 4mm bore stopcock plug, see 89062-774. For replacement 6mm bore stopcock plug, see 89062-776.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty