Would you like to log in again?
Bovine Desoxyribonuclease I (from Pancreas)
Supplier: Thermo Scientific
J60027-EXR J60027-EXU J60027-EXU J60027-EXR
AAJ60027-EXREA 82.4 USD InStock
CAAAJ60027-EXR AAJ60027-EXR CAAAJ60027-EXU AAJ60027-EXU
Bovine Desoxyribonuclease I (from Pancreas)
Enzymes
MW = 31 kDa.
Endonuclease that splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty