Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ingenol-3-angelate ≥98%

Supplier: Thermo Scientific Chemicals

J60601-MCR J60601-MCR
AAJ60601-MCREA 320.59 USD InStock
CAAAJ60601-MCR AAJ60601-MCR
Ingenol-3-angelate ≥98%
Ingenol-3-angelate
Protein kinase C activator


Formula: C₂₅H₃₄O₆
MW: 430.541 g/mol
Storage Temperature: Freezer
MDL Number: MFCD07784504
CAS Number: 75567-37-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty