Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ionomycin calcium salt 99%, crystallised

Supplier: Thermo Scientific Chemicals
Warning

J60628-LB0 J60628-LB0
AAJ60628-LB0EA 275.04 USD InStock
AAJ60628-LB0 CAAAJ60628-LB0
Ionomycin calcium salt 99%, crystallised
Ionomycin calcium salt
Crystallized powder

Highly selective and potent calcium ionophore


Formula: C₄₁H₇₀CaO₉
Storage Temperature: Freezer
MDL Number: MFCD00151183
CAS Number: 56092-82-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty