Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calphostin C ≥95%

Supplier: Thermo Scientific Chemicals

J60647-EXG J60647-EXG
AAJ60647-EXGEA 249.57 USD InStock
CAAAJ60647-EXG AAJ60647-EXG
Calphostin C ≥95%
Calphostin C
Potent inhibitor of protein kinase C


Formula: C₄₄H₃₈O₁₄
Storage Temperature: Ambient
MDL Number: MFCD00133155
CAS Number: 121263-19-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty