Would you like to log in again?
Calf Alkaline phosphatase (from Intestine)
Supplier: Thermo Scientific
J61037-LB0 J61037-MCR J61037-LB0 J61037-MCR
AAJ61037-MCREA 184.6 USD InStock
AAJ61037-MCR AAJ61037-LB0 CAAAJ61037-MCR CAAAJ61037-LB0
Calf Alkaline phosphatase (from Intestine)
Enzymes
MW = 140 kDa
Used to dephosphorylate proteins; also used to dephosphorylate the 5' termini of DNA or RNA to prevent self ligation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty