Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rabbit Native Actin (from Muscle)

Supplier: Thermo Scientific
J61490-MCR J61490-MCR
AAJ61490-MCREA 147.78 USD InStock
CAAAJ61490-MCR AAJ61490-MCR
Rabbit Native Actin (from Muscle)
Proteins and Peptides
Lyophilized powder

CAS No.: 51005-14-2
Storage Temperature: -30°C to -10°C
Physical Form: Lyophilized powder
Appearance: White to off-white
MDL No.: MFCD00130338

Used in cell transport studies, and inhibits deoxyribonuclease I

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty