Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human beta-Endorphin

Supplier: Thermo Scientific
J62374-MCR J62374-MCR
AAJ62374-MCREA 225.17 USD InStock
AAJ62374-MCR CAAAJ62374-MCR
Human beta-Endorphin
Proteins and Peptides
CAS No.: 61214-51-5
Molecular Formula: C158H251N39O46S
Formula Weight: 3465.06
Physical Form: Powder
Appearance: White

An endogenous opioid peptide neurotransmitter found in the neurons of both the central and peripheral nervous systemy

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty