Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Okadaic acid sodium salt 98%, Powder

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 9,10-deepithio-9,10-didehydroacanthifolicin sodium saltOA sodium salt

J62463-EXS
AAJ62463-EXSEA 587.43 USD InStock
AAJ62463-EXS
Okadaic acid sodium salt 98%, Powder
Okadaic acid sodium salt
Potent inhibitor of serine/threonine-specific protein phosphatases 1 and 2A (PP1 and PP2A). Does not inhibit tyrosine or alkaline phosphatases


Formula: C₄₄H₆₇NaO₁₃
MW: 827.00 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00210208
CAS Number: 209266-80-8
UN: 3462
ADR: 6.1,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty