Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LB Agar Plate with Tetracycline, Thermo Scientific

Supplier: Thermo Scientific
J62894-EQF J62894-EQF
AAJ62894-EQFEA 151.14 USD InStock
CAAAJ62894-EQF AAJ62894-EQF
LB Agar Plate with Tetracycline, Thermo Scientific
Media for Microbiology Prepared Media for Microbiology
This LB agar plate contains tetracycline at a concentration of 10 µg/mL.

Storage Temperature: 2°C to 8°C
Physical Form: Solid

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty