Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Ethoxyresorufin

Supplier: Thermo Scientific Chemicals
Synonyms: Resorufin ethyl ether

J62987-LB0 J62987-LB0
AAJ62987-LB0EA 338 USD InStock
CAAAJ62987-LB0 AAJ62987-LB0
7-Ethoxyresorufin
7-Ethoxyresorufin
Fluorimetric substrate for detection of cytochrome P450 linked enzymes


Formula: C₁₄H₁₁NO₃
MW: 241.246 g/mol
Melting Pt: 237…240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037661
CAS Number: 5725-91-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty