Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chymostatin

Supplier: Thermo Scientific
J63275-LB0 J63275-LB0
AAJ63275-LB0EA 140.99 USD InStock
CAAAJ63275-LB0 AAJ63275-LB0
Chymostatin
Proteins and Peptides
CAS No.: 9076-44-2
Molecular Formula: C31H41N7O6
Formula Weight: 604.90
Physical Form: Powder
Appearance: White to off-white
Melting Point: 276-278°
MDL No.: MFCD00071059

A highly specific chymotrypsin inhibitor produced by actinomycetes

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty