Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Human Glucagon (19-29)

Supplier: Thermo Scientific
J63443-MCR J63443-LB0 J63443-LB0 J63443-MCR
AAJ63443-MCREA 66.24 USD InStock
CAAAJ63443-LB0 AAJ63443-MCR CAAAJ63443-MCR AAJ63443-LB0
Human Glucagon (19-29)
Proteins and Peptides
CAS No.: 64790-15-4
Molecular Formula: C61H89N15O18S
Formula Weight: 1352.50
Physical Form: Powder
Appearance: White

This fragment inhibits calcium activated ATPase activity

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty