Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Salmon Calcitonin

Supplier: Thermo Scientific
J63531-EXD J63531-EXD
AAJ63531-EXDEA 162.28 USD InStock
CAAAJ63531-EXD AAJ63531-EXD
Salmon Calcitonin
Proteins and Peptides
CAS No.: 47931-85-1
Molecular Formula: C145H240N44O48S2
Formula Weight: 3417.90
Physical Form: Powder
Appearance: White
Solubility: Soluble in water at 1mg/ml
MDL No.: MFCD00133859

A peptide hormone

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty