Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

FURA-2 pentasodium salt fluorescent indicator

Supplier: Thermo Scientific Chemicals

J63686-MCR J63686-MCR
AAJ63686-MCREA 162.29 USD InStock
CAAAJ63686-MCR AAJ63686-MCR
FURA-2 pentasodium salt fluorescent indicator
FURA-2 pentasodium salt
A very sensitive fluorescent calcium chelator


Formula: C₂₉H₂₂N₃Na₅O₁₄
Storage Temperature: Freezer
MDL Number: MFCD00083328
CAS Number: 96314-98-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty