Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LB Agar Plate with Spectinomycin, Thermo Scientific

Supplier: Thermo Scientific
J63732-EQF J60541-EQF J60541-EQF J63732-EQF
AAJ60541-EQFEA 200.97 USD InStock
CAAAJ60541-EQF AAJ60541-EQF AAJ63732-EQF CAAAJ63732-EQF
LB Agar Plate with Spectinomycin, Thermo Scientific
Media for Microbiology Prepared Media for Microbiology
LB Agar plate with spectinomycin is used in the resistance of 16S ribosomal RNA in Pasteurella multocida.

Storage Temperature: 2°C to 8°C
Physical Form: Solid

Caution: Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty