Synonyms:
(2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-Ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-((2S,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl)-8-oxononadeca-2,10,12,16,18-pentaenoic acid
J63784-EXG
AAJ63784-EXGEA
367.5
USD
InStock
AAJ63784-EXG
Leptomycin B 1 mM in ethanol ≥99%
Leptomycin B
This leptomycin B is a potent, specific inhibitor of nuclear export signal (NES)-dependent protein export from the nucleus.
Purity: 99+%
CAS No.: 87081-35-4
Molecular Formula: C33H48O6
Formula Weight: 540.73
Storage Temperature: -30°C to -10°C
Flash Point: 11°(52°F)
Physical Form: Liquid
Appearance: Clear colorless
Odor: Alcohol-like
Merck Reference: 14,5444
UN No.: 1993
MDL No.: MFCD06795848
Potent, specific inhibitor of nuclear export signal (NES)-dependent protein export from the nucleus