Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alamethicin

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: U-22324Antibiotic U-22324

J63829-MCR J63829-LB0
AAJ63829-MCREA 168.65 USD InStock
AAJ63829-MCR AAJ63829-LB0
Alamethicin
Alamethicin
Alamethicin is used as a channel-forming ionophore that activates membrane enzymes, membrane channel-forming peptide.

  • White powder

This alamethicin is a 20-amino acid channel-forming peptide antibiotic that can function as a monovalent cation ionophore. Alamethicin has been used to develop methods for the routine assessment of potential new drug candidates to elicit their pharmacokinetic drug interactions.

A channel-forming ionophore that activates membrane enzymes

Soluble in DMSO and ethanol


Formula: C₉₂H₁₅₀N₂₂O₂₅
MW: 1964.308 g/mol
Melting Pt: 267…270 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151517
CAS Number: 59588-86-2
UN: 3462
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty