Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pig Dynorphin A (1-13)

Supplier: Thermo Scientific
J64015-MCR J64015-LB0 J64015-LB0 J64015-MCR
AAJ64015-LB0EA 231.28 USD InStock
CAAAJ64015-MCR AAJ64015-LB0 AAJ64015-MCR CAAAJ64015-LB0
Pig Dynorphin A (1-13)
Proteins and Peptides
CAS No.: 72957-38-1
Molecular Formula: C75H126N24O15
Formula Weight: 1603.95
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00076366

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty