Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Salmon LH-RH

Supplier: Thermo Scientific
J64173-MCR J64173-MCR
AAJ64173-MCREA 60.86 USD InStock
CAAAJ64173-MCR AAJ64173-MCR
Salmon LH-RH
Proteins and Peptides
CAS No.: 86073-88-3
Molecular Formula: C60H73N15O13
Formula Weight: 1212.31
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
MDL No.: MFCD00133497

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty