Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Alpha-Melanocyte Stimulating Hormone amide

Supplier: Thermo Scientific
J64310-LB0 J64310-LB0 J64310-MCR J64310-MCR
AAJ64310-MCREA 65.41 USD InStock
AAJ64310-MCR CAAAJ64310-LB0 CAAAJ64310-MCR AAJ64310-LB0
Alpha-Melanocyte Stimulating Hormone amide
Proteins and Peptides
Molecular Formula: C77H109N21O19S
Formula Weight: 1664.88
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty