Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Laminin (925-933)

Supplier: Thermo Scientific
J64340-EXD J64340-EXD J64340-MCR J64340-MCR
AAJ64340-MCREA 151.14 USD InStock
AAJ64340-MCR CAAAJ64340-MCR AAJ64340-EXD CAAAJ64340-EXD
Laminin (925-933)
Proteins and Peptides
CAS No.: 110590-60-8
Molecular Formula: C40H62N12O14S
Formula Weight: 967.05
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to off-white
MDL No.: MFCD00076474

Derived from residues 925-933 of human laminin B1, this peptide binds the laminin receptor

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty