Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Caspase-1 Inhibitor IV

Supplier: Thermo Scientific
J64583-MCR J64583-MCR
AAJ64583-MCREA 163.34 USD InStock
CAAAJ64583-MCR AAJ64583-MCR
Caspase-1 Inhibitor IV
Proteins and Peptides
CAS No.: 154674-81-4
Molecular Formula: C33H42N4O10
Formula Weight: 654.71
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Soluble in DMSO
MDL No.: MFCD00798794

A competitive, cell-permeable, and irreversible inhibitor of caspases -1, -4, and -5

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty