Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Leu-Leu-Glu-7-amino-4-methylcoumarin

Supplier: Thermo Scientific Chemicals
Synonyms: Z-Leu-Leu-Glu-AMC

J64613-LB0 J64613-LB0
AAJ64613-LB0EA 148.11 USD InStock
AAJ64613-LB0 CAAAJ64613-LB0
Z-Leu-Leu-Glu-7-amino-4-methylcoumarin
Z-Leu-Leu-Glu-7-amino-4-methylcoumarin
A fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (caspase-like) activity of the proteasome which may be stimulated in the presence of Mg2+ ions and rapidly inactivated by N-acetylimidazole


Formula: C₃₅H₄₄N₄O₉
MW: 664.756 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01074989
CAS Number: 348086-66-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty