Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Calpain Inhibitor XII, solid

Supplier: Thermo Scientific Chemicals

J64751-LB0 J64751-MCR J64751-LB0 J64751-MCR
AAJ64751-MCREA 116.66 USD InStock
CAAAJ64751-LB0 AAJ64751-MCR CAAAJ64751-MCR AAJ64751-LB0
Calpain Inhibitor XII, solid
Calpain Inhibitor XII
  • Appearnace: Pale yellow solid

A cell-permeable dipeptidyl -ketoamide compound that acts as a potent, selective, reversible, and active site inhibitor of calpain-1


Formula: C₂₆H₃₄N₄O₅
MW: 482.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06411395
CAS Number: 181769-57-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty