Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Z-Gly-Gly-Leu-7-Amino-4-Methylcoumarin

Supplier: Thermo Scientific
J64796-LB0 J64796-LB0
AAJ64796-LB0EA 87.14 USD InStock
AAJ64796-LB0 CAAAJ64796-LB0
Z-Gly-Gly-Leu-7-Amino-4-Methylcoumarin
Proteins and Peptides
Molecular Formula: C28H32N4O7
Formula Weight: 536.57
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to pale yellow
MDL No.: MFCD00152029

  • Light Sensitive

A fluorogenic substrate for the chymotrypsin-like activity of the proteasome

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty