Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kisspeptin-10

Supplier: Thermo Scientific
J65487-MCR J65487-MCR
AAJ65487-MCREA 111.59 USD InStock
CAAAJ65487-MCR AAJ65487-MCR
Kisspeptin-10
Proteins and Peptides
Molecular Formula: C63H83N17O14
Formula Weight: 1302.44
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White to off-white
Solubility: Soluble in water at 1mg/ml
MDL No.: MFCD03452696

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty