Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Pen Vibration Meter, Extech
Supplier: Extech
VB400
89370-190EA 1167.71 USD InStock
89370-190
Pen Vibration Meter, Extech
Vibration Meters
Pen Vibration Meter that measures RMS Acceleration and Velocity with metric and imperial units of measure
This meter features Zero adjustment, Data Hold, and Low battery indicator functions. It is also Water and dust resistant to IP65.
Certifications: CE marked.
Ordering information: Test pin tip, magnetic base, 4 AA batteries, and pouch case included. 1 year warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty