Would you like to log in again?
Stirrer Blades, PTFE, for Button-Type Shafts, Ace Glass Incorporated
Supplier: Ace Glass
8082-06 8082-100 8082-04 8082-08 8082-02
89062-058EA 28.14 USD InStock
89062-058 89062-060 89062-062 89062-054 89062-056
Stirrer Blades, PTFE, for Button-Type Shafts, Ace Glass Incorporated
Stirrers Stirring Blades
Oval-shaped PTFE stirrer blades are designed for use with 10mm O.D
button-type stirring shafts.
Ordering information: Item 89062-062 is supplied as a pack of 6 blades, with two each of blades 89062-054 and 89062-056, and one each of blades 89062-058 and 89062-060.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty