Would you like to log in again?
Reduced End Tube Filters, Ace Glass Incorporated
Supplier: Ace Glass
7212-14 7212-22 7212-28 7212-18 7212-08 7212-04
89058-480EA 124.89 USD InStock
89058-480 89058-398 89058-422 89058-318 89058-428 89058-290
Reduced End Tube Filters, Ace Glass Incorporated
Filters Filter Tubes
Reduced end tube filters are supplied in a range of sizes, from a 20 mm disc size to the 50 mm disc size.
The smaller sizes are used to remove end products of a reaction during recirculation of liquid reactants. The larger units are suitable for filtering operations in the preparation of organic chemicals and biologic solutions. All sizes are supplied with hose connection ends. Fabricated from borosilicate glass.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty