Would you like to log in again?
Ammonia Salicylate Reagent Powder Pillows, Hach
Supplier: Hach
2653299 2653299
89231-158PK 235.98 USD InStock
CA26532-99 89231-158
Ammonia Salicylate Reagent Powder Pillows, Hach
Environmental Test Kits
For determination of Ammonia Nitrogen by the Salicylate method
Hach Method 8155.
Ordering information: Pack of 100 Powder Pillows.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty