Would you like to log in again?
Nitrification Inhibitor for BOD, Formula 2533™, TCMP, Hach
Supplier: Hach
Formula 2533™
253334 253334
97026-568EA 518.07 USD InStock
97026-568 CA2533-34
Nitrification Inhibitor for BOD, Formula 2533™, TCMP, Hach
Standards Environmental Analysis Standards
TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate
Use 0.16 gram in a 300-mL sample to inhibit nitrogenous oxygen demand in the BOD test.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty