Would you like to log in again?
HQ440d Benchtop Meter Package with LBOD101 Dissolved Oxygen Probe for BOD Measurement, Hach
Supplier: Hach
8508500 8508500
89233-214EA 7963.34 USD InStock
89233-214 CA89233-214
HQ440d Benchtop Meter Package with LBOD101 Dissolved Oxygen Probe for BOD Measurement, Hach
Electrochemical Meters Oxygen Meters
Designed for water quality experts, the Hach HQ440d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • Make your laboratory testing more efficient with a flexible meter to measure critical water quality parametersimprove productivity and save laboratory space with the HQd benchtop meter
  • All benchtop HQd meters provide an intuitive user interface for simple operation and accurate results
  • Trust your measurements with the IntelliCAL™ BOD smart probe that stores all calibrations in the probe
  • Enhance productivity with an ultrabright screen and large font size for easy to read results
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous laboratory use. HQd meters feature a simple user interface that does not require manuals or training to operate. The breakthrough luminescent BOD probe has no membranes to replace for hassle-free maintenance and stabilizes quickly LDO technology and integrated stirrer.
Ordering information: Meter package includes HQ440d Benchtop Meter, LBOD101 BOD probe, 100 disposable BOD bottles, bottle stoppers (qty 25), snap-over bottle caps (qty 24), 4 AA batteries, power adapter, USB cable for data transfer, quick-start guide, user manual, and documentation CD.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty