Would you like to log in again?
Angiotensin I (1-9) Trifluoroacetate
Supplier: Bachem
4033395.0100 4033395.0025
H-5038.0025BAEA 304.81 USD InStock
H-5038.0025BA H-5038.0100BA
Angiotensin I (1-9) Trifluoroacetate
Proteins and Peptides
The sequences given for angiotensin I (H-1680) and II (H-1705) correspond to the sequences of human, horse, sheep, pig, mouse, and rat angiotensins I and II.
Angiotensin I (1-9) is found in vivo and accumulates in animals treated with ACE inhibitors. It has been shown that the nonapeptide may be generated in heart and kidney by endothelium-associated ACE2. Kramkowski et al. observed that DRVYIHPFH generated from Ang I in platelets enhances thrombosis in rats.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty