Would you like to log in again?
Silica Reagent Set, High Range, 10 ml, Hach
Supplier: Hach
2429600 2429600
89231-386EA 220.25 USD InStock
89231-386 CA2429600
Silica Reagent Set, High Range, 10 ml, Hach
Environmental Test Kits
For High Range Silica determination by the Silicomolybdate method
Hach Method 8185. Range: 1 - 100 mg/L SiO2.
Ordering information: Set contains Acid Reagent Powder Pillows, Citric Acid Powder Pillows and Molybdate Reagent Powder Pillows for 100 tests.
Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty