Would you like to log in again?
Fluoride Ionic Strength Adjustor (ISA) Powder Pillows, Hach
Supplier: Hach
258999 258999
97021-786PK 156.22 USD InStock
CA97021-786 97021-786
Fluoride Ionic Strength Adjustor (ISA) Powder Pillows, Hach
Environmental Test Kits
Fluoride Total Ionic Strength Adjustor Buffer (TISAB) for use when making fluoride determinations by electrode methods
Eliminates interferences from aluminum and iron. Powder form does not dilute samples. Ideal for determining very low fluoride levels.
Ordering information: Pack of 100 Powder Pillows.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty