Would you like to log in again?
Acid Reagent Powder Pillows for HR Silica, Hach
Supplier: Hach
2107469 2107469
89231-310PK 84.94 USD InStock
89231-310 CA21074-69
Acid Reagent Powder Pillows for HR Silica, Hach
Environmental Test Kits
For determination of high range silica by the silicomolybdate method (Hach method 8185) Range: 1-100.0 mg/L SiO2
Also used in the iodometric method (Hach method 10100) for the determination of high range hypchlorite (50-150 g/L as Cl2). Pack of 100 powder pillows.
Ordering information: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty