Would you like to log in again?
Total Hardness Test Kit, Model HA-71A, Hach
Supplier: Hach
145201 145201
89231-236EA 192.29 USD InStock
CA11024-936 89231-236
Total Hardness Test Kit, Model HA-71A, Hach
Environmental Test Kits
Water hardness is caused almost entirely by calcium and magnesium ion.
  • Utilizes drop count titration a simple, economical titration method.
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field.
Other di- and trivalent metals have a similar effect, but usually are not present in high enough concentration in potable waters to cause problems. Hardness increases soap consumption in laundries and causes scale in boilers. With the Model HA-71A test kit, simply count the number of drops required to change the solution color to determine hardness concentration. Utilizing drop count titration, this kit provides dual ranges and contains approximately 100 tests.
Ordering information: Hazardous Items: Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty