Would you like to log in again?
PFA Evaporation Dish, Savillex
Supplier: Savillex
700-150
30617-528EA 95.88 USD InStock
30617-528
PFA Evaporation Dish, Savillex
Trays/Dishes/Pots Evaporating Basins/Dishes
PFA evaporation dish with flat bottom and molded pour spout
Service temperature range is –200 to 260°C (–328 to 500°F).
Flat bottomed for efficinet heating using a hot plate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty