Would you like to log in again?
Wash Bottle, PFA, Savillex
Supplier: Savillex
700-500
30617-534EA 639.64 USD InStock
30617-534
Wash Bottle, PFA, Savillex
Bottles Wash Bottles
This 500 ml PFA wash bottle is manufactured using Savillex's proprietary molding technology, which produces an extremely smooth surface.
The spout, closure, and bottle are all made from the highest purity PFA resin, making the wash bottle suitable for the most demanding ultratrace applications. Ideal for use with ICP-MS.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty