Would you like to log in again?
PFA Pipette Tips, Savillex
Supplier: Savillex
700-600
30617-536EA 54.58 USD InStock
30617-536
PFA Pipette Tips, Savillex
Pipette Tips
These PFA pipette tips are made from the highest purity grade PFA resin and are designed for the most demanding ultratrace metals analysis applications
For a constant supply of high purity acid, use the Savillex DST-1000 Acid Purification System (30621-038).
The 8mm opening fits most popular 1000µL and adjustable pipettes. Orifice size is 8mm. Designed to be reused after cleaning in ultrapure acid.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty