Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HQ440d Benchtop Meter Package with ISEF121 Fluoride ISE Electrode, Hach
Supplier: Hach
8508000 8508000
89174-064EA 7664.49 USD InStock
89174-064 CA89174-064
HQ440d Benchtop Meter Package with ISEF121 Fluoride ISE Electrode, Hach
Electrochemical Meters Ion Meters
Designed for water quality experts, the HQ440d benchtop meter is an advanced laboratory meter that takes the guesswork out of measurements.
  • Flexible benchtop meter makes your lab testing of critical water quality parameters more efficient
  • Intuitive user interface for simple operation and accurate results
  • Trust your measurements IntelliCAL™ smart probes store all calibrations in the probe
  • Enhance productivity with an ultrabright screen and large font size for easy to read results
  • Convenient kit includes everything you need to start testing
HQd meters connect with smart probes that automatically recognize the testing parameter, calibration history, and method settings to minimize errors and setup time. The advanced benchtop meter is designed to withstand years of continuous lab use. HQd meters feature a simple user interface that does not require manuals or training to operate.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty