Would you like to log in again?
Silica Reagent Set, Ultra Low Range Rapid Liquid™, Hach
Supplier: Hach
2678500 2678500
89231-166EA 301.8 USD InStock
89231-166 CA11024-472
Silica Reagent Set, Ultra Low Range Rapid Liquid™, Hach
Environmental Test Kits
For determination of ultra-low range Silica by the Heteropoly Blue Rapid Liquid™ method
  • Convenience
  • Performance
  • Efficiency
Hach method 8282. Range: 3 - 1000 μg/L.
Ordering information: Set includes: Molybdate 3 Reagent Solution, Citric Acid F Reagent Solution, Amino Acid Reagent Dilution Solvent and Amino Acid F Reagent Powder for approximately 250 tests. Requires Pour-Thru Cell.
Hazardous Items:
Items with this mark may be considered hazardous under some shipping conditions.If necessary, we will change your selected shipping method to accommodate these items.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty